Learn More
CD45RO Mouse anti-Human, PE, Clone: UCHL1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 347967
Description
CD45RO, clone UCHL-1, is derived from hybridization of mouse P3/NS-1/1-Ag4-1 myeloma cells with spleen cells from BALB/c mice immunized with an interleukin-2 (IL-2)-dependent human T-cell line.
CD45RO, a member of the CD45 family that includes CD45, CD45RA, and CD45RB, recognizes a 180-kilodalton (kd) isoform of the leucocyte common antigen (LCA). The CD45 antigen is a protein tyrosine phosphatase.
Specifications
| CD45RO | |
| Monoclonal | |
| 6.25μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human IL-2-dependent T-cell line | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| UCHL1 | |
| PE | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2a κ |