missing translation for 'onlineSavingsMsg'
Learn More

CD48 Mouse anti-Human, PE, Clone: TÜ145, BD

Product Code. 15866878
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15866878 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15866878 Supplier BD Biosciences Supplier No. 552855

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The TÜ145 monoclonal antibody specifically binds to CD48. CD48 belongs to the CD2 family within the immunoglobulin superfamily. CD48 is a ∽43 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein. CD48 is strongly expressed on lymphocytes, monocytes and macrophages and weakly on granulocytes. The expression of CD48 is upregulated following lymphocyte activation and by Epstein-Barr virus (EBV) infection. CD48 binds with high affinity to CD244 and with low affinity to CD2. These receptor-ligand interactions can augment or inhibit leukocyte responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD48
Applications Flow Cytometry
Classification Monoclonal
Clone TÜ145
Conjugate PE
Description BCM1; BLAST; BLAST1; Hulym3; MEM-102; OX45; SLAMF2; TCT.1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.