Learn More
CD49f Rat anti-Human, Alexa Fluor 647, Clone: GoH3, BD
Rat Monoclonal Antibody
€ 71.14 - € 357.00
Specifications
| Antigen | CD49f |
|---|---|
| Clone | GoH3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15801979
|
BD Biosciences
562494 |
25 Tests |
€ 71.70 € 71.14 / Each Save € 0.56 0.78% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15891949
|
BD Biosciences
562473 |
100 Tests |
€ 357.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GoH3 monoclonal antibody specifically binds to CD49f which is also known as integrin α6 chain. CD49f is a ∽150 kDa type I transmembrane glycoprotein that belongs to the integrin alpha chain family of extracellular matrix and cell adhesion receptors. The integrin α6 subunit associates with the integrin β1 chain (CD29) to form VLA-6 and with the integrin β4 chain (CD104) to form the integrin α6β4 complex, also known as the laminin and kalinin receptor. CD49f is expressed mainly on T cells, monocytes, platelets, epithelial cells, endothelial cells, perineural cells, and trophoblasts of placenta. GoH3 recognizes an extracellular epitope of integrin α6 on human, mouse and bovine cells. GoH3 has been reported to block the binding of integrin α6 to laminin P1 and E8 fragments.
Specifications
| CD49f | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified | |
| ITGA6; IThe GoH3 monoclonal ntegrin alpha-6; Integrin α6 chain; VLA-6; ITA6 |
| GoH3 | |
| Monoclonal | |
| Rat | |
| Human | |
| Mouse mammary tumor cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.