Learn More
CD4a Mouse, FITC, Clone: 74-12-4, BD
Description
The 74-12-4 (also known as clone PT4) monoclonal antibody specifically binds to CD4, a 55-kDa antigen expressed on T lymphocytes. This antibody does not react with CTL effectors, CTL precursors, or NK cells (ie, CD8[bright] cells) and it does not cross-react with human or bovine cells. Two peripheral T-helper lymphocyte phenotypes can be distinguished in the pig: CD4+CD8- and CD4+CD8[dull]. mAb 74-12-4 has been reported to inhibit proliferative responses of peripheral blood lymphocytes to mitogen, soluble antigen, and alloantigen. It is only marginally effective for in vivo depletion of peripheral CD4+ T cells. Two alloantigenic forms of CD4 have been recognized in miniature swine based upon their recognition (CD4.1) or lack of recognition (CD4.2) by mAb 74-12-4; the CD4.2 phenotype displays an autosomal recessive, non-MHC-linked, pattern of inheritance. The molecular basis for the polymorphism is a cluster of nucleotide differences leading to multiple amino-acid substitutions in the Ig CDR2-like loop structure. This mAb was clustered as anti-CD4a at the First International Swine CD Workshop. It has been reported to crossreact with chicken leukocytes.
Specifications
Specifications
| Antigen | CD4a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 74-12-4 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | dd miniature swine thymocytes |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.