Learn More
CD51/CD61 Mouse anti-Human, FITC, Clone: 23C6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555505
Description
The 23C6 monoclonal antibody specifically binds to the Integrin αvβ3 heterodimeric complex (CD51/CD61) that is expressed on osteoclasts, endothelial cells, macrophages, melanoma cells, some B cells and in very low amounts on platelets. This antibody has been reported to crossreact with chicken and rabbit, but not with rat αvβ3. The integrin αvβ3 heterodimer, often referred to as the vitronectin receptor, binds several ligands in addition to vitronectin including, von Willebrand factor, thrombospondin, osteopontin, bone sialoprotein, neural adhesion molecule L1, laminin, fibronectin, and fibrinogen. The binding of some ligands to the vitronectin receptor has been reported to be inhibited by the 23C6 monoclonal antibody.
Specifications
| CD51/CD61 | |
| Monoclonal | |
| FITC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Bovine, Human, Rabbit | |
| IgG1 κ |
| Flow Cytometry | |
| 23C6 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.