missing translation for 'onlineSavingsMsg'
Learn More

CD56 Mouse anti-Human, PE-Cy7, Clone: B159, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 557747

 View more versions of this product

Product Code. 15869028

  • € 455.00 / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD56
Monoclonal
PE-Cyanine7
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human NK Cells
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
B159
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.