Learn More
CD79a Mouse anti-Human, APC, Clone: HM47, BD
Beschreibung
The HM47 monoclonal antibody specifically binds to the cytoplasmic domain of CD79a. CD79a is a 47 kDa type 1 transmembrane glycoprotein present on B lymphocytes. CD79a is also referred to as mb-1, IGA and Ig-alpha (Ig α ). It is expressed on B cells at various stages of differentiation, from pre-B cell stage, probably before expression of cytoplasmic µ chain, to plasma cell stage, in which it is detected only in the cytoplasm. CD79a associates with CD79b to form part of the B-cell receptor complex. It has been suggested that CD79a may play a role in mediating the transport of IgM to the cell surface. This antibody has been found to crossreact with permeabilized A20 cells (mouse B cell line).
Spezifikation
Spezifikation
| Antigen | CD79a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HM47 |
| Conjugate | APC |
| Description | CD79a, immunoglobulin-associated alpha ; CD79A; Ig-alpha; IGA; Igα; MB-1 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human CD79a Peptide |
| Purification Method | Affinity Purified |
| Mehr anzeigen |
For Research Use Only.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.