Learn More
CD8 Mouse anti-Human, FITC, Clone: RPA-T8, BD
Mouse Monoclonal Antibody
€ 107.86 - € 491.13
Specifications
| Antigen | CD8 |
|---|---|
| Clone | RPA-T8 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15821499
|
BD Biosciences
561947 |
25 Tests |
€ 109.00 € 107.86 / Each Save € 1.14 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15831499
|
BD Biosciences
561948 |
500 Tests |
€ 493.00 € 491.13 / Each Save € 1.87 0.38% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RPA-T8 monoclonal antibody specifically binds to CD8 alpha (CD8α) . CD8α is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily. CD8α is expressed by the majority of thymocytes, by subpopulations of αβ T cells and γδ T cells and by some NK cells. Cell surface CD8α is expressed either as a disulfide-linked homodimer (CD8αα) αβ T cells coexpress both CD8αα homodimers and CD8αβ heterodimers whereas some γδ T cells and NK cells express CD8αα homodimers. CD8 plays important roles in T cell activation and selection. The extracellular IgSF domain of CD8 α binds to a non-polymorphic determinant on HLA class I molecules (α3 domain) and enables CD8 to function as a co-receptor with MHC class I-restricted TCR during T cell recognition of antigen. The cytoplasmic domain of CD8α associates with Lck, a Src family protein tyrosine kinase that is involved in intracellular signaling. The RPA-T8 and HIT8a monoclonal antibodies are not cross-blocking. This clone has been reported to react with a subset of peripheral blood lymphocytes, but not monocytes nor granuloyctes, of baboon and both rhesus and cynomolgus macaque monkey. In general, a higher frequency of CD8+ and CD4+CD8+ lymphocytes are observed in non-human primates compared to normal human donors.
Spécification
| CD8 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 |
| RPA-T8 | |
| Monoclonal | |
| Mouse | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| Human CD8a | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.