Learn More
CD86 Mouse anti-Rat, PE, Clone: 24F, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551396
Description
The 24F antibody reacts with CD86 (B7-2), a member of the Ig superfamily of transmembrane proteins. CD86, a ligand for CD28 and CD152 (CTLA-4), is one of the accessory molecules that plays an important role in T cell-B cell costimulatory interactions. CD86 is predominantly expressed on antigen-presenting cells. It can be upregulated on splenic B cells by in vitro polyclonal activation, such as LPS treatment. 24F mAb is reported to block the costimulatory function of rat CD86 and to immunoprecipitate CD86 from Lewis-S1 cell lysates. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 24F
Isotype: IgG1 κ
Species Reactivity: Rat
Immunogen: HTLV-1-transformed rat T-cell line Lewis-S1
Flow Cytometry
Specifications
| CD86 | |
| Monoclonal | |
| 0.2mg/mL | |
| B7-2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 24F | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| HTLV-1-transformed rat T-cell line Lewis-S1 | |
| 0.1 mg | |
| Immunology | |
| Rat | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.