Learn More
CD87 Mouse anti-Human, PE, Clone: VIM5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555768
Description
Reacts with the urokinase plasminogen activator receptor (uPA-R), a 50-65kDa single chain glycosylphosphatidylinositol (GPI)-linked glycoprotein. uPA-R was clustered as CD87 in the Fifth Human Leukocyte Differentiation Antigens Workshop. It is expressed on neutrophils, monocytes, tissue macrophages, endothelial cells, and activated T cells. CD87 plays a role in cell-associated fibrinolytic activity and tissue invasion. This antibody may be useful in the study of myeloid cell differentiation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: VIM5
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD87 | |
| Monoclonal | |
| PE | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| VIM5 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.