Learn More
CD94 Mouse anti-Human, Unlabeled, Clone: HP-3D9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555887
Description
Reacts with a 70kDa disulfide-linked dimer, also known as Kp43, expressed on natural killer (NK) cells. It is also expressed on γ/δ TCR+ T lymphocytes and on some CD8+CD56+ α/β TCR+ clones. Reports demonstrate that HP-3D9 markedly inhibits cytolytic activity of polyclonally-activated NK cells. CD94 (Kp43) plays a role in regulation of the function of NK cell activation and adhesion
Host Species: Mouse
Clone: HP-3D9
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
| CD94 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human NK Cells | |
| 1 mL | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| HP-3D9 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.