Learn More
CDC42 effector protein (Rho GTPase binding) 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CDC42EP3.
Brand: Abnova H00010602-A01.50uL
Description
CDC42, a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to, and negatively regulate the function of, CDC42. This protein can interact with CDC42, as well as with the ras homolog gene family, member Q (ARHQ/TC10). Expression of this protein in fibroblasts has been shown to induce pseudopodia formation. [provided by RefSeq
Sequence: IHIGKEGQHDVFGDISFLQGNYELLPGNQEKAHLGQFPGHNEFFRANSTSDSVFTETPSPVLKNAISSpecifications
| CDC42 effector protein (Rho GTPase binding) 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CDC42EP3. | |
| CDC42EP3 | |
| BORG2/CEP3/FLJ46903/UB1 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 10602 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006449 | |
| CDC42EP3 | |
| CDC42EP3 (NP_006440, 42 a.a. ∼ 108 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.