missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cell division cycle associated 3, Mouse, Clone: 3B9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CDCA3.
Brand: Abnova H00083461-M11.100ug
This item is not returnable.
View return policy
Description
Sequence: MGSAKSVPVTPARPPPHNKHLARVADPRSPSAGILRTPIQVESSPQPGLPAGEQLEGLKHAQDSDPRSPTLGIARTPMKTSSGDPPSPLVKQLSEVFETEDSKSNLPPEPVLPPEAPLSSELDLPLGTQLSVEEQMPPWNQTEFPSKQVFSKEEARQPTETPVASQSSDKPSRDPETPRSSGSMRNRWKPNSSKVLGRSPLTILQDDNSPGTLTLRQGKRPSPLSENVSELKEGAILGTGRLLKTGGRAWEQGQDHDKENQHFPLVESSpecifications
| cell division cycle associated 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC002551 | |
| CDCA3 | |
| CDCA3 (AAH02551, 1 a.a. ∼ 268 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3B9 | |
| Mouse monoclonal antibody raised against a full length recombinant CDCA3. | |
| CDCA3 | |
| GRCC8/MGC2577/TOME-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 83461 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction