Learn More
CDH4, Mouse, Clone: 2E2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CDH4.
Brand: Abnova H00001002-M01.100ug
Description
This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Based on studies in chicken and mouse, this cadherin is thought to play an important role during brain segmentation and neuronal outgrowth. In addition, a role in kidney and muscle development is indicated. Of particular interest are studies showing stable cis-heterodimers of cadherins 2 and 4 in cotransfected cell lines. Previously thought to interact in an exclusively homophilic manner, this is the first evidence of cadherin heterodimerization. [provided by RefSeq
Sequence: AADADVDPNIGPYVFELPFVPAAVRKNWTITRLNGDYAQLSLRILYLEAGMYDVPIIVTDSGNPPLSNTSIIKVKVCPCDDNGDCTTIGAVAAAGLGTGASpecifications
| CDH4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001794 | |
| CDH4 | |
| CDH4 (NP_001785, 635 a.a. ∼ 734 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2E2 | |
| Mouse monoclonal antibody raised against a partial recombinant CDH4. | |
| CDH4 | |
| CAD4/FLJ22202/FLJ40547/MGC126700/MGC138355/RCAD | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1002 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.