Learn More
cyclin-dependent kinase 7, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CDK7 protein.
Brand: Abnova H00001022-D01P.100ug
Description
The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle. [provided by RefSeq
Sequence: MALDVKSRAKRYEKLDFLGEGQFATVYKARDKNTNQIVAIKKIKLGHRSEAKDGINRTALREIKLLQELSHPNIIGLLDAFGHKSNISLVFDFMETDLEVIIKDNSLVLTPSHIKAYMLMTLQGLEYLHQHWILHRDLKPNNLLLDENGVLKLADFGLAKSFGSPNRAYTHQVVTRWYRAPELLFGARMYGVGVDMWAVGCILAELLLRVPFLPGDSDLDQLTRIFETLGTPTEEQWPDMCSLPDYVTFKSFPGIPLHHIFSAAGDDLLDLIQGLFLFNPCARITATQALKMKYFSNRPGPTPGCQLPRPNCPVETLKEQSNPALAIKRKRTEALEQGGLPKKLIFSpecifications
| cyclin-dependent kinase 7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human CDK7 protein. | |
| CDK7 | |
| CAK1/CDKN7/MO15/STK1/p39MO15 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| In situ PLA, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001799.2 | |
| CDK7 | |
| CDK7 (NP_001790.1, 1 a.a. ∼ 346 a.a) full-length human protein. | |
| 100 μg | |
| Cell Cycle | |
| 1022 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.