Learn More
CDKN2D, Mouse, Clone: 2E10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CDKN2D.
Brand: Abnova H00001032-M08.100ug
Description
The protein encoded by this gene is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to form a stable complex with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. The abundance of the transcript of this gene was found to oscillate in a cell-cycle dependent manner with the lowest expression at mid G1 and a maximal expression during S phase. The negative regulation of the cell cycle involved in this protein was shown to participate in repressing neuronal proliferation, as well as spermatogenesis. Two alternatively spliced variants of this gene, which encode an identical protein, have been reported. [provided by RefSeq
Sequence: MLLEEVRAGDRLSGAAARGDVQEVRRLLHRELVHPDALNRFGKTALQVMMFGSTAIALELLKQGVSPNVQDTSGTSPVHDAARTGFLDTLKVLVEHGADVNVPDGTGALPIHLAVQEGHTAVVSFLAAESDLHRRDARGLTPLELALQRGAQDLVDILQGHMVAPLSpecifications
| CDKN2D | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC001822 | |
| CDKN2D | |
| CDKN2D (AAH01822, 1 a.a. ∼ 166 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 2E10 | |
| Mouse monoclonal antibody raised against a full-length recombinant CDKN2D. | |
| CDKN2D | |
| INK4D/p19/p19-INK4D | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1032 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.