missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDV3 homolog (mouse), Mouse, MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CDV3 protein.
Brand: Abnova H00055573-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MAETEERSLDNFFAKRDKKKKKERSNRAASAAGAAGSAGGSSGAAGAAGGGAGAGTRPGDGGTASAGAAGPGAATKAVTKDEDEWKELEQKEVDYSGLRVQAMQISSEKEEDDNEKRQDPGDNWEEGGGGGGGMEKSSGPWNKTAPVQAPPAPVIVTETPEPAMTSGVYRPPGARLTTTRKTPQGPPEIYSDTQFPSLQSTAKHVESRNRYLKSpecifications
| CDV3 homolog (mouse) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human CDV3 protein. | |
| CDV3 | |
| H41 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC007338 | |
| CDV3 | |
| CDV3 (AAH07338, 1 a.a. ∼ 213 a.a) full-length human protein. | |
| RUO | |
| 55573 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction