Learn More
CDX1, Mouse, Clone: 1E9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CDX1.
Brand: Abnova H00001044-M18.100ug
Description
This gene is a member of the caudal-related homeobox transcription factor gene family. The encoded DNA-binding protein regulates intestine-specific gene expression and enterocyte differentiation. It has been shown to induce expression of the intestinal alkaline phosphatase gene, and inhibit beta-catenin/T-cell factor transcriptional activity. [provided by RefSeq
Sequence: MYVGYVLDKDSPVYPGPARPASLGLGPANYGPPAPPPAPPQYPDFSSYSHVEPAPAPPTAWGAPFPAPKDDWAAAYGPGPAAPAASPASLAFGPPPDFSPVPAPPGPGPGLLAQPLGGPGTPSSPGAQRPTPYEWMRRS*Specifications
| CDX1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001804 | |
| CDX1 | |
| CDX1 (NP_001795, 1 a.a. ∼ 139 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1E9 | |
| Mouse monoclonal antibody raised against a full length recombinant CDX1. | |
| CDX1 | |
| MGC116915 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1044 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.