missing translation for 'onlineSavingsMsg'
Learn More
Learn More
centrosomal protein 57kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CEP57 protein.
Brand: Abnova H00009702-D01P.100ug
This item is not returnable.
View return policy
Description
Translokin binds basic fibroblast growth factor (FGF2; MIM 134920) and mediates its nuclear translocation and mitogenic activity (Bossard et al., 2003 [PubMed 12717444]).[supplied by OMIM
Sequence: MAAASVSAASGSHLSNSFAEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQELTSQLLAAENKCNLLEKQLEYMRNMIKHAEMERTSVLEKQVSLERERQHDQTHVQSQLEKLDLLEQEYNKLTTMQALAEKKMQELEAKLHEEEQERKRMQAKAAELQTGLETNRLIFEDKATPCVPNARRIKKKKSKPPEKKSSRNYFGAQPHYRLCLGDMPFVAGKSTSPSHAVVANVQLVLHLMKQHSKALCNDRVINSIPLAKQVSSRGGKSKKLSVTPPSSNGINEELSEVLQTLQDEFGQMSFDHQQLAKLIQESPTVELKDKLECELEALVGRMEAKANQITKVRKYQAQLEKQKLEKQKKELKATKKTLDEERNSSSRSGITGTTNKKDFMKLRPGEKRRKNLQLLKDMQSIQNSLQSSSLCWDYSpecifications
| centrosomal protein 57kDa | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human CEP57 protein. | |
| CEP57 | |
| KIAA0092/PIG8/TSP57 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9702 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014679.3 | |
| CEP57 | |
| CEP57 (NP_055494.2, 1 a.a. ∼ 500 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction