Learn More
CHML, Mouse, Clone: 5G4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CHML.
Brand: Abnova H00001122-M03.100ug
Description
The product of the CHML gene supports geranylgeranylation of most Rab proteins and may substitute for REP-1 in tissues other than retina. CHML is localized close to the gene for Usher syndrome type II. [provided by RefSeq
Sequence: IGEESTVVWQDLIHETEEAITLRKKDETIQHTEAFCYASQDMEDNVEEIGALQKNPSLGVSNTFTEVLDSALPEESQLSYFNSDEMPAKHTQKSDTEISLEVTDVEESVESpecifications
| CHML | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001821 | |
| CHML | |
| CHML (NP_001812, 68 a.a. ∼ 177 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 5G4 | |
| Mouse monoclonal antibody raised against a partial recombinant CHML. | |
| CHML | |
| FLJ10071/FLJ13361/REP2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1122 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.