Learn More
chromatin modifying protein 1A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CHMP1A protein.
Brand: Abnova H00005119-D01P.100ug
Description
This gene encodes a member of the CHMP/Chmp family of proteins which are involved in multivesicular body sorting of proteins to the interiors of lysosomes. The initial prediction of the protein sequence encoded by this gene suggested that the encoded protein was a metallopeptidase. The nomenclature has been updated recently to reflect the correct biological function of this encoded protein. [provided by RefSeq
Sequence: MDDTLFQLKFTAKQLEKLAKKAEKDSKAEQAKVKKALLQKNVECARVYAENAIRKKNEGVNWLRMASRVDAVASKVQTAVTMKGVTKNMAQVTKALDKALSTMDLQKVSSVMDRFEQQVQNLDVHTSVMEDSMSSATTLTTPQEQVDSLIMQIAEENGLEVLDQLSQLPEGASAVGESSVRSQEDQLSRRLAALRNSpecifications
| chromatin modifying protein 1A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human CHMP1A protein. | |
| CHMP1A | |
| CHMP1/KIAA0047/PCOLN3/PRSM1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5119 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002768.2 | |
| CHMP1A | |
| CHMP1A (NP_002759.2, 1 a.a. ∼ 196 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.