missing translation for 'onlineSavingsMsg'
Learn More

chimerin (chimaerin) 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CHN1 protein.

Brand:  Abnova H00001123-B01.50uL

Product Code. 16173191

  • € 349.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes GTPase-activating protein for p21-rac and a phorbol ester receptor. It plays an important role in ocular motor axon pathfinding. Heterozygous missense mutations in this gene cause Duane's retraction syndrome 2 (DURS2). Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MALTLFDTDEYRPPVWKSYLYQLQQEAPHPRRITCTCEVENRPKYYGREFHGMISREAADQLLIVAEGSYLIRESQRQPGTYTLALRFGSQTRNFRLYYDGKHFVGEKRFESIHDLVTDGLITLYIETKAAEYIAKMTINPIYEHVGYTTLNREPAYKKHMPVLKETHDERDSTGQDGVSEKRLTSLVRRATLKENEQIPKYEKIHNFKVHTFRGPHWCEYCANFMWGLIAQGVKCADCGLNVHKQCSKMVPNDCKPDLKHVKKVYSCDLTTLVKAHTTKRPMVVDMCIREIESRGLNSEGLYRVSGFSDLIEDVKMAFDRDGEKADISVNMYEDINIITGALKLYFRDLPIPLITYDAYPKFIESAKIMDPDEQLETLHEALKLLPPAHCETLRYLMAHLKRVTLHEKENLMNAENLGIVFGPTLMRSPELDAMAALNDIRYQRLVVELLIKNEDILF
Specifications

Specifications

chimerin (chimaerin) 1
Polyclonal
Mouse polyclonal antibody raised against a full-length human CHN1 protein.
CHN1
ARHGAP2/CHN/DURS2/RHOGAP2
Mouse
50 μL
Membrane Receptors
Primary
Human
Serum
Western Blot
Unconjugated
No additive
NM_001822
CHN1
CHN1 (NP_001813, 1 a.a. ∼ 459 a.a) full-length human protein.
RUO
Yes
1123
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.