Learn More
carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CHST2.
Brand: Abnova H00009435-A01.50uL
Description
N-acetylglucosamine-6-O-sulfotransferases, such as CHST2, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) to position 6 of a nonreducing N-acetylglucosamine (GlcNAc) residue (Uchimura et al., 1998 [PubMed 9722682]).[supplied by OMIM
Sequence: DPVKTLRRVYDFVGLLVSPEMEQFALNMTSGSGSSSKPFVVSARNATQAANAWRTALTFQQIKQVEEFCYQPMAVLGYERVNSPEEVKDLSKTLLRKPRLSpecifications
| carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CHST2. | |
| CHST2 | |
| C6ST/GST2 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 9435 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004267 | |
| CHST2 | |
| CHST2 (NP_004258, 431 a.a. ∼ 530 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.