Learn More
CIDEC, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CIDEC protein.
Brand: Abnova H00063924-B01P.50ug
Description
DNA fragmentation factor (DFF) induces the fragmentation of DNA associated with apoptosis. A novel family of cell death-inducing DFF45 (MIM 601882)-like effectors (CIDEs), including CIDEC, can also promote apoptosis (Liang et al., 2003 [PubMed 12429024]).[supplied by OMIM
Sequence: MEYAMKSLSLLYPKSLSRHVSVRTSVVTQQLLSEPSPKAPRARPCRVSTADRSVRKGIMAYSLEDLLLKVRDTLMLADKPFFLVLEEDGTTVETEEYFQALAGDTVFMVLQKGQKWQPPSEQGTRHPLSLSHKPAKKIDVARVTFDLYKLNPQDFIGRLNVKATFYDTYSLSYDLHCCGAKRIMKEAFRWALFSMQATGHVLLGTSCYLQQLLDATEEGQPPKGKASSLIPTCLKILQSpecifications
| CIDEC | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human CIDEC protein. | |
| CIDEC | |
| CIDE-3/FLJ20871/Fsp27 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 63924 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC016851 | |
| CIDEC | |
| CIDEC (AAH16851, 1 a.a. ∼ 238 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.