Learn More
CKS2, Mouse, Clone: 1G8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CKS2.
Brand: Abnova H00001164-M03.100ug
Description
CKS2 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS2 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects specialized role for the encoded protein. [provided by RefSeq
Sequence: MAHKQIYYSDKYFDEHYEYRHVMLPRELSKQVPKTHLMSEEEWRRLGVQQSLGWVHYMIHEPEPHILLFRRPLPKDQQKSpecifications
| CKS2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001827 | |
| CKS2 | |
| CKS2 (NP_001818, 1 a.a. ∼ 79 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunofluorescence, Western Blot | |
| 1G8 | |
| Mouse monoclonal antibody raised against a partial recombinant CKS2. | |
| CKS2 | |
| CKSHS2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1164 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.