Learn More
Claudin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532353
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is small intestine tissue.
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of a network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different claudin composition.Specifications
Claudin 3 | |
Polyclonal | |
CLDN3 | |
C7orf1, CPE-R2, CPETR2, HRVP1, RVP1, CPE-R 2, CPE-receptor 2, Clostridium perfringens enterotoxin receptor 2, claudin-3, ventral prostate.1 protein homolog, ventral prostate.1-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1365, 65130, 12739 | |
-20°C or -80°C if preferred | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Q63400, Q9Z0G9, O15551 | |
CLDN3 | |
Synthetic peptide derived from the C-terminus of mouse Claudin-3 | |
500 μL | |
Primary | |
Chicken, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |