missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKSR family member 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CNKSR3 protein.
Brand: Abnova H00154043-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MEPVTKWSPKQVVDWTRGLDDCLQQYVHKFEREKINGEQLLQISHQDLEELGVTRIGHQELVLEAVDLLCALNYGLETDNMKNLVLKLRASSHNLQNYISSRRKSPAYDGNTSRKAPNEFLTSVVELIGAAKALLAWLDRAPFTGITDFSVTKNKIIQLCLDLTTTVQKDCFVAEMEDKVLTVVKVLNGICDKTIRSTTDPVMSQCACLEEVHLPNIKPGEGLGMYIKSTYDGLHVITGTTENSPADRSQKIHAGDEVIQVNQQTVVGWQLKNLVKKLRENPTGVVLLLKKRPTGSFNFTPAPLKNLRWKPPLVQTSPPPATTQSPESTMDTSLKKEKSAILDLYIPPPPAVPYSPRDENGSFVYGGSSKCKQPLPGPKGSESPNSFLDQESRRRRFTIADSDQLPGYSVETNILPTKMREKTPSYGKPRPLSMPADGNWMGIVDPFARPRGHGRKGEDALCRYFSNERIPPIIEESSSPPYRFSRPTTERHLVRGADYIRGSRCYINSDLHSSATIPFQEEGTKKKSGSSATKSSSTEPSLLVSWFTRLKLLTHSpecifications
| CNKSR family member 3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human CNKSR3 protein. | |
| CNKSR3 | |
| FLJ31349/MAGI1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_173515 | |
| CNKSR3 | |
| CNKSR3 (NP_775786.2, 1 a.a. ∼ 555 a.a) full-length human protein. | |
| 100 μg | |
| Neurobiology | |
| 154043 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction