Learn More
coatomer protein complex, subunit beta 1, Mouse, Clone: 3E10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant COPB.
Brand: Abnova H00001315-M08.100ug
Description
This gene encodes a protein subunit of the coatomer complex associated with nonclathrin coated vesicles. The coatomer complex, also known as the coat protein complex 1, forms in the cytoplasm and is recruited to the Golgi by activated guanosine triphosphatases. Once at the Golgi membrane, the coatomer complex may assist in the movement of protein and lipid components back to the endoplasmic reticulum. Alternatively spliced transcript variants have been described. [provided by RefSeq
Sequence: TVNTNMVDLNDYLQHILKSTNMKCLTPEKALSGYCGFMAANLYARSIFGEDALANVSIEKPIHQGPDAAVTGHIRIRAKSQGMALSLGDKINLSQKKTSISpecifications
| coatomer protein complex, subunit beta 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_016451 | |
| COPB1 | |
| COPB (NP_057535, 854 a.a. ∼ 953 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 1315 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
| ELISA | |
| 3E10 | |
| Mouse monoclonal antibody raised against a partial recombinant COPB. | |
| COPB1 | |
| COPB/DKFZp761K102/FLJ10341 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.