Learn More
COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant COPS3.
Brand: Abnova H00008533-A01.50uL
Description
The protein encoded by this gene possesses kinase activity that phosphorylates regulators involved in signal transduction. It phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation. The gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq
Sequence: SRVQLSGPQEAEKYVLHMIEDGEIFASINQKDGMVSFHDNPEKYNNPAMLHNIDQEMLKCIELDERLKAMDQEITVNPQFVQKSMGSQEDDSGNKPSSYSpecifications
| COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant COPS3. | |
| COPS3 | |
| CSN3/SGN3 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 8533 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003653 | |
| COPS3 | |
| COPS3 (NP_003644, 324 a.a. ∼ 422 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.