missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coronin 1B Polyclonal Antibody

Product Code. 13485277
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13485277 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13485277 Supplier Invitrogen™ Supplier No. PA530775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (99%), Rat (97%), Rabbit (98%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin 1B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Coro1b
Gene Accession No. Q9BR76, Q9WUM3
Gene Alias cor-1; Coro1b; coronin 1b; coronin 2; coronin, actin binding protein 1B; coronin, actin binding protein, 1B; coronin, actin binding protein, 1B (coronin 2); coronin, actin-binding protein, 1B; coronin-1B; coronin-1B-like; CORONIN-2; Coronin-like protein pp66; coroninse
Gene Symbols Coro1b
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Coronin 1B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23789, 57175
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.