Learn More
CSNK2A1, Mouse, Clone: 3D9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CSNK2A1.
Brand: Abnova H00001457-M01.100ug
Description
Casein kinase II is a serine/threonine protein kinase that phosphorylates acidic proteins such as casein. The kinase exists as a tetramer and is composed of an alpha, an alpha-prime, and two beta subunits. The alpha subunits contain the catalytic activity while the beta subunits undergo autophosphorylation. The protein encoded by this gene represents the alpha subunit. While this gene is found on chromosome 20, a related transcribed pseudogene is found on chromosome 11. Three transcript variants encoding two different proteins have been found for this gene. [provided by RefSeq
Sequence: MSGPVPSRARVYTDVNTHRPREYWDYESHVVEWGNQDDYQLVRKLGRGKYSEVFEAINITNNEKVVVKILKPVKKKKIKREIKILENLRGGPNIITLADISpecifications
| CSNK2A1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC011668 | |
| CSNK2A1 | |
| CSNK2A1 (AAH11668, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, In situ PLA, Western Blot | |
| 3D9 | |
| Mouse monoclonal antibody raised against a partial recombinant CSNK2A1. | |
| CSNK2A1 | |
| CK2A1/CKII | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1457 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.