missing translation for 'onlineSavingsMsg'
Learn More
Learn More
chondroitin sulfate proteoglycan 5 (neuroglycan C), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CSPG5.
Brand: Abnova H00010675-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: KKLYLLKTENTKLRRTNKFRTPSELHNDNFSLSTIAEGSHPNDDPSAPHKIQEVLKSCLKEEESFNIQNSMSPKLEGGKGDQADLDVNCLQNNLTSpecifications
| chondroitin sulfate proteoglycan 5 (neuroglycan C) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CSPG5. | |
| CSPG5 | |
| MGC44034/NGC | |
| Mouse | |
| 50 μL | |
| Neurobiology | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006574 | |
| CSPG5 | |
| CSPG5 (NP_006565, 445 a.a. ∼ 539 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 10675 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction