Learn More
cysteine and glycine-rich protein 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CSRP1.
Brand: Abnova H00001465-A01.50uL
Description
This gene encodes a member of the cysteine-rich protein (CSRP) family. This gene family includes a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this gene product occurs in proteins with critical functions in gene regulation, cell growth, and somatic differentiation. [provided by RefSeq
Sequence: EAPGHRPTTNPNASKFAQKIGGSERCPRCSQAVYAAEKVIGAGKSWHKACFRCAKCGKGLESTTLADKDGEIYCKGCYAKNFGPKGFGFGQGAGALVHSSpecifications
| cysteine and glycine-rich protein 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CSRP1. | |
| CSRP1 | |
| CRP/CRP1/CSRP/CYRP/D1S181E/DKFZp686M148 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 1465 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004078 | |
| CSRP1 | |
| CSRP1 (NP_004069, 94 a.a. ∼ 192 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.