Learn More
cullin 4B, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Description
This gene is a member of the cullin family. The encoded protein forms a complex that functions as an E3 ubiquitin ligase and catalyzes the polyubiquitination of specific protein substrates in the cell. The protein interacts with a ring finger protein, and is required for the proteolysis of several regulators of DNA replication including chromatin licensing and DNA replication factor 1 and cyclin E. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
| Antigen | cullin 4B |
| Applications | Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human CUL4B protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CUL4B |
| Gene Accession No. | NM_003588 |
| Gene Alias | DKFZp686F1470/KIAA0695/MRXHF2/MRXSC/SFM2 |
| Gene Symbols | CUL4B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.