Learn More
CYP2J2, Mouse, Clone: 2D10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CYP2J2.
Brand: Abnova H00001573-M01.100ug
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is thought to be the predominant enzyme responsible for epoxidation of endogenous arachidonic acid in cardiac tissue. [provided by RefSeq
Sequence: LHRDPTEWATPDTFNPDHFLENGQFKKREAFMPFSIGKRACLGEQLARTELFIFFTSLMQKFTFRPPNNEKLSLKFRMGITISPVSHRLCASpecifications
| CYP2J2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000775 | |
| CYP2J2 | |
| CYP2J2 (NP_000766, 408 a.a. ∼ 498 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2D10 | |
| Mouse monoclonal antibody raised against a partial recombinant CYP2J2. | |
| CYP2J2 | |
| CPJ2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1573 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.