Learn More
Cytokeratin 14, 15, 16 and 19 Set Mouse anti-Human, PE, Clone: KA4, BD
Description
Reacts with cytokeratins 14, 15, 16 and 19 present in the cytoplasm of a wide variety of human simple epithelium. Human epithelium has been shown to contain cytoplasmic filaments of the cytokeratin type. There have been 19 different polypeptides identified and they demonstrate a characteristic pattern for each type of epithelium. This reagent is useful for intracellular staining for flow cytometric analysis, western blot and immunohistochemical staining of both, acetone-fixed frozen tissue sections and formalin-fixed paraffin embedded tissue sections with citrate pretreatment.
Specifications
Specifications
| Antigen | Cytokeratin 14, 15, 16 and 19 Set |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | KA4 |
| Conjugate | PE |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.