Learn More
DDT, Mouse, Clone: 1D5, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant DDT.
Brand: Abnova H00001652-M04.100ug
Description
D-dopachrome tautomerase converts D-dopachrome into 5,6-dihydroxyindole. The DDT gene is related to the migration inhibitory factor (MIF) in terms of sequence, enzyme activity, and gene structure. DDT and MIF are closely linked on chromosome 22. [provided by RefSeq
Sequence: MPFLELDTNLPANRVPAGLEKRLCAAAASILGKPADRVNVTVRPGLAMALSGSTEPCAQLSISSIGVVGTAEDNRSHSAHFFEFLTKELALGQDRILIRFFPLESWQIGKIGTVMTFLSpecifications
| DDT | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC015508 | |
| DDT | |
| DDT (AAH15508, 1 a.a. ∼ 118 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunoprecipitation | |
| 1D5 | |
| Mouse monoclonal antibody raised against a full-length recombinant DDT. | |
| DDT | |
| DDCT | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1652 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.