Learn More
DGKA, Mouse, Clone: 2B7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant DGKA.
Brand: Abnova H00001606-M02.100ug
Description
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It acts as a modulator that competes with protein kinase C for the second messenger diacylglycerol in intracellular signaling pathways. It also plays an important role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. [provided by RefSeq
Sequence: MAKERGLISPSDFAQLQKYMEYSTKKVSDVLKLFEDGEMAKYVQGDAIGYEGFQQFLKIYLEVDNVPRHLSLALFQSFETGHCLNETNVTKDVVCLNDVSCYFSLLEGGRPEDKLEFTFKLYDTDRNGILDSSEVDKIILQMMRVAEYLDWDVSELRPILQEMMKEIDYDGSGSVSQAEWVRAGATTVPLLVLLGLEMTLKDDGQHMWRPKRFPRPVYCNLCESSIGLGKQGLSCNLCKYTVHDQCAMKALPCEVSTYAKSRKDIGVQSHVWVRGGCESGRCDRCQKKIRIYHSLTGLHCVWCHLEIHDDCLQAVGHECDCGLLRDHILPPSSIYPSVLASGPDRKNSKTSQKTMDDLNLSTSEALRIDPVPNTHPLLVFVNPKSGGKQGQRVLWKFQYILNPRQVFNLLKDGPEIGLRLFKDVPDSRILVCGGDGTVGWILETIDKANLPVLPPVAVLPLGTGNDLARCLRWGGGYEGQNLAKILKDLEMSKVVHMDRWSVEVIPQQTEEKSDPVPFQIINNYFSIGVDASIAHRFYIMREKYPEKFNSRMKNKLWYFEFATSESIFSTCKKLEESLTVEICGKPLDLSNLSLEGIAVLNIPSMHGGSNLWGDTRRPHGDIYGINQALGATAKVITDPDILKTCVPDLSDKRLEVVGLEGAIEMGQIYTKLKNAGRRLAKCSEITFHTTKTLPMQIDGEPWVQTPCTIKITHKNQMPMLMGPPPRSTNFFGFLSSpecifications
| DGKA | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC031870 | |
| DGKA | |
| DGKA (AAH31870, 1 a.a. ∼ 735 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunohistochemistry (PFA fixed), In situ PLA | |
| 2B7 | |
| Mouse monoclonal antibody raised against a full-length recombinant DGKA. | |
| DGKA | |
| DAGK/DAGK1/DGK-alpha/MGC12821/MGC42356 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1606 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Safety and Handling
- DGKA monoclonal antibody (M02), clone 2B7
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium chloride, sodium phosphate dibasic, potassium phosphate monobasic, potassium chloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.