missing translation for 'onlineSavingsMsg'
Learn More

dehydrogenase/reductase (SDR family) member 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16165768
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16165768 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16165768 Supplier Abnova Supplier No. H00009249B01P.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human DHRS3 protein.

Short-chain dehydrogenases/reductases (SDRs), such as DHRS3, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM

Sequence: MVWKRLGALVMFPLQMIYLVVKAAVGLVLPAKLRDLSRENVLITGGGRGIGRQLAREFAERGARKIVLWGRTEKCLKETTEEIRQMGTECHYFICDVGNREEVYQTAKAVREKVGDITILVNNAAVVHGKSLMDSDDDALLKSQHINTLGQFWTTKAFLPRMLELQNGHIVCLNSVLALSAIPGAIDYCTSKASAFAFMESLTLGLLDCPGVSATTVLPFHTSTEMFQGMRVRFPNLFPPLKPETVARRTVEAVQLNQALLLLPWTMHALVILKSILPQAALEEIHKFSGTYTCMNTFKGRT

Specifications

Antigen dehydrogenase/reductase (SDR family) member 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DHRS3 protein.
Formulation PBS with no preservative; pH 7.4
Gene DHRS3
Gene Accession No. NM_004753.4
Gene Alias RDH17/Rsdr1/SDR1/SDR16C1/retSDR1
Gene Symbols DHRS3
Host Species Mouse
Immunogen DHRS3 (NP_004744.2, 1 a.a. ∼ 302 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9249
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.