Learn More
discs, large homolog 4 (Drosophila), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant DLG4.
Brand: Abnova H00001742-A01.50uL
Description
This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with another MAGUK protein, DLG2, and is recruited into NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: QGKHCILDVSANAVRRLQAAHLHPIAIFIRPRSLENVLEINKRITEEQARKAFDRATKLEQEFTECFSAIVEGDSFEEIYHKVKRVIEDLSGPYIWVPARERSpecifications
| discs, large homolog 4 (Drosophila) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant DLG4. | |
| DLG4 | |
| FLJ97752/FLJ98574/PSD95/SAP-90/SAP90 | |
| Mouse | |
| 50 μL | |
| Neurobiology | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001365 | |
| DLG4 | |
| DLG4 (NP_001356, 665 a.a. ∼ 766 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 1742 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.