Learn More
distal-less homeobox 4, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human DLX4 protein.
Brand: Abnova H00001748-D01P.100ug
Description
Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. Three transcript variants have been described for this gene, however, the full length nature of one variant has not been described. Studies of the two splice variants revealed that one encoded isoform functions as a repressor of the beta-globin gene while the other isoform lacks that function. [provided by RefSeq
Sequence: MTSLPCPLPGRDASKAVFPDLAPVPSVAAAYPLGLSPTTAASPNLSYSRPYGHLLSYPYTEPANPGDSYLSCQQPAALSQPLCGPAEHPQELEADSEKPRLSPEPSERRPQAPAKKLRKPRTIYSSLQLQHLNQRFQHTQYLALPERAQLAAQLGLTQTQVKIWFQNKRSKYKKLLKQNSGGQEGDFPGRTFSVSPCSPPLPSLWDLPKAGTLPTSGYGNSFGAWYQHHSSDVLASPQMMSpecifications
| distal-less homeobox 4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human DLX4 protein. | |
| DLX4 | |
| BP1/DLX7/DLX8/DLX9 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_138281.1 | |
| DLX4 | |
| DLX4 (NP_612138.1, 1 a.a. ∼ 240 a.a) full-length human protein. | |
| 100 μg | |
| Transcription Regulation | |
| 1748 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.