Learn More
aspartyl aminopeptidase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human DNPEP protein.
Brand: Abnova H00023549-D01P.100ug
Description
The protein encoded by this gene is an aminopeptidase which prefers acidic amino acids, and specifically favors aspartic acid over glutamic acid. It is thought to be a cytosolic protein involved in general metabolism of intracellular proteins. [provided by RefSeq
Sequence: MTRNSSTIIAFAVGGQYVPGNGFSLIGAHTDSPCLRVKRRSRRSQVGFQQVGVETYGGGIWSTWFDRDLTLAGRVIVKCPTSGRLEQQLVHVERPILRIPHLAIHLQRNINENFGPNTEMHLVPILATAIQEELEKGTPEPGLSMLWMSGTIRSSCPCSVPIWGSpecifications
| aspartyl aminopeptidase | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human DNPEP protein. | |
| DNPEP | |
| ASPEP/DAP | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC003040 | |
| DNPEP | |
| DNPEP (AAH03040.1, 1 a.a. ∼ 164 a.a) full-length human protein. | |
| 100 μg | |
| Stem Cell Biology | |
| 23549 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.