Learn More
DPP4, Mouse, Clone: 1C5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DPP4.
Brand: Abnova H00001803-M03.100ug
Description
The protein encoded by this gene is identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. [provided by RefSeq
Sequence: GWVGRFRPSEPHFTLDGNSFYKIISNEEGYRHICYFQIDKKDCTFITKGTWEVIGIEALTSDYLYYISNEYKGMPGGRNLYKIQLSDYTKVTCLSRELNPSpecifications
| DPP4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC065265 | |
| DPP4 | |
| DPP4 (AAH65265, 352 a.a. ∼ 451 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 1C5 | |
| Mouse monoclonal antibody raised against a partial recombinant DPP4. | |
| DPP4 | |
| ADABP/ADCP2/CD26/DPPIV/TP103 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1803 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.