Learn More
ectodysplasin A receptor, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EDAR.
Brand: Abnova H00010913-A01.50uL
Description
This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia. [provided by RefSeq
Sequence: TKDEDYGCVPCPAEKFSKGGYQICRRHKDCEGFFRATVLTPGDMENDAECGPCLPGYYMLENRPRNIYGMVCYSCLLAPPNTKECVGATSGASANFPGTSGSSTLSPFQHAHKELSpecifications
| ectodysplasin A receptor | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EDAR. | |
| EDAR | |
| DL/ED1R/ED3/ED5/EDA-A1R/EDA1R/EDA3/FLJ94390 | |
| Mouse | |
| 50 μL | |
| Apoptosis | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_022336 | |
| EDAR | |
| EDAR (NP_071731, 64 a.a. ∼ 178 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 10913 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.