Learn More
EEF1B2, Mouse, Clone: 3A5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EEF1B2.
Brand: Abnova H00001933-M10.100ug
Description
This gene encodes a translation elongation factor. The protein is a guanine nucleotide exchange factor involved in the transfer of aminoacylated tRNAs to the ribosome. Alternative splicing results in three transcript variants which differ only in the 5' UTR. [provided by RefSeq]
Sequence: VPSQADVAVFEAVSSPPPADLCHALRWYNHIKSYEKEKASLPGVKKALGKYGPADVEDTTGSGSpecifications
| EEF1B2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001959.3 | |
| EEF1B2 | |
| EEF1B2 (NP_001950.1, 29 a.a. ∼ 91 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3A5 | |
| Mouse monoclonal antibody raised against a partial recombinant EEF1B2. | |
| EEF1B2 | |
| EEF1B/EEF1B1/EF1B | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1933 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.