Learn More
epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Description
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq
Specifications
Specifications
| Antigen | epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian) |
| Applications | In situ PLA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human EGFR protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | EGFR |
| Gene Accession No. | NM_201283 |
| Gene Alias | ERBB/ERBB1/HER1/PIG61/mENA |
| Gene Symbols | EGFR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.