missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EH-domain containing 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EHD3.
Brand: Abnova H00030845-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: KRMQDQLQAQDFSKFQPLKSKLLEVVDDMLAHDIAQLMVLVRQEESQRPISpecifications
| EH-domain containing 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EHD3. | |
| EHD3 | |
| PAST3 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 30845 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_014600 | |
| EHD3 | |
| EHD3 (NP_055415, 357 a.a. ∼ 406 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction