missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eukaryotic translation initiation factor 2-alpha kinase 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EIF2AK3.
Brand: Abnova H00009451-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: KWQEKMDEIWLKDESTDWPLSSPSPMDAPSVKIRRMDPFSTKEHIEIIAPSPQRSRSFSVGISCDQTSSSESQFSPLEFSGMDHEDISESVDAAYNLQDSSpecifications
| eukaryotic translation initiation factor 2-alpha kinase 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EIF2AK3. | |
| EIF2AK3 | |
| DKFZp781H1925/HRI/PEK/PERK/WRS | |
| Mouse | |
| 50 μL | |
| Kinases | |
| Primary | |
| Human, Rat | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004836 | |
| EIF2AK3 | |
| EIF2AK3 (NP_002437, 665 a.a. ∼ 764 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 9451 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction