Learn More
eukaryotic translation initiation factor 4E nuclear import factor 1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EIF4ENIF1.
Brand: Abnova H00056478-A01.50uL
Description
The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predominantly cytoplasmic; its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: NPRPGTPLHLAMVQQQLQRSVLHPPGSGSHAAAVSVQTTPQNVPSRSGLPHMHSQLEHRPSQRSSSPVGLAKWFGSDVLQQPLPSMPAKVISVDELEYRQSpecifications
| eukaryotic translation initiation factor 4E nuclear import factor 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EIF4ENIF1. | |
| EIF4ENIF1 | |
| 4E-T/Clast4/FLJ21601/FLJ26551 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 56478 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_019843 | |
| EIF4ENIF1 | |
| EIF4ENIF1 (NP_062817, 886 a.a. ∼ 985 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.