missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF5, Mouse, Clone: 1D9-4B9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant EIF5.
Brand: Abnova H00001983-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MSVNVNRSVSDQFYRYKMPRLIAKVEGKGNGIKTVIVNMVDVAKALNRPPTYPTKYFGCELGAQTQFDVKNDRYIVNGSHEANKLQDMLDGFIKKFVLCPECENPETDLHVNPKKQTIGNSCKACGYRGMLDTHHKLCTFILKNPPENSDSGTGKKEKEKKNRKGKDKENGSVSSSETPPPPPPPNEINPPPHTMEEEEDDDWGEDTTEEAQRRRMDEISDHAKVLTLSDDLERTIEERVNILFDFVKKKKEEGVIDSSDKEIVAEAERLDVKAMGPLVLTEVLFNEKIREQIKKYRRHFLRFCHNNKKAKRYLLHGLECVVAMHQAQLISKIPHILKEMYDADLLEEEVIISWSEKASKKYVSKELAKEIRVKAEPFIKWLKEAEEESSGGEEEDEDENIEVVYSKAASVPKVETVKSDNKDDDIDIDAISpecifications
| EIF5 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC032866 | |
| EIF5 | |
| EIF5 (AAH32866, 1 a.a. ∼ 431 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1D9-4B9 | |
| Mouse monoclonal antibody raised against a full-length recombinant EIF5. | |
| EIF5 | |
| EIF-5/EIF-5A | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1983 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction